missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ADAM9 Ectodomain Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB949
This item is not returnable.
View return policy
Description
ADAM9 Monoclonal specifically detects ADAM9 in Mouse samples. It is validated for Western Blot.
Specifications
| ADAM9 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| a disintegrin and metalloproteinase domain 9 (meltrin gamma), ADAM 9, ADAM metallopeptidase domain 9, ADAM metallopeptidase domain 9 (meltrin gamma), Cellular disintegrin-related protein, cone rod dystrophy 9, CORD9, disintegrin and metalloproteinase domain-containing protein 9, EC 3.4.24, EC 3.4.24.-, MCMP, MCMPMDC9KIAA0021Mltng, MDC9, Meltrin gamma, Meltrin-gamma, Metalloprotease/disintegrin/cysteine-rich protein 9, MLTNG, Myeloma cell metalloproteinase | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8754 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 122823 | |
| Western Blot 1 ug/mL | |
| Q61072 | |
| ADAM9 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ADAM9 Ala205-Cys673 Accession # Q61072 | |
| 500 μg | |
| Primary | |
| Detects mouse ADAM9 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) ADAM9 is observed and no cross-reactivity with rhADAM8, recombinant mouse ADAM10, rhADAM15, rhBACE, or rhTACE is observed. In Western blots, approximately 100% cross-reactivity with rhADAM9 and rmADAM15 is observed and less than 5% cross-reactivity with rmADAM10 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction