missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ADAM33 Ectodomain Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2434-SP
This item is not returnable.
View return policy
Description
ADAM33 Monoclonal specifically detects ADAM33 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ADAM33 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q923W9 | |
| ADAM33 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ADAM33 Ectodomain Glu205-Ala702 Accession # Q923W9 | |
| 25 μg | |
| Primary | |
| Detects mouse ADAM33 Ectodomain in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse ADAM9, 10, 15, recombinant human (rh) ADAM8, rhTACE/ADAM17, rhADAM19, or rhADAM33 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 219515 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| a disintegrin and metalloprotease 33, a disintegrin and metalloproteinase domain 33, ADAM 33, ADAM metallopeptidase domain 33, C20orf153, chromosome 20 open reading frame 153, disintegrin and metalloproteinase domain-containing protein 33, disintegrin and reprolysin metalloproteinase family protein, DJ964F7.1, DKFZp434K0521, EC 3.4.24.-, FLJ35308, FLJ36751, MGC149823, MGC71889 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 80332 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction