missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ADAM10 Ectodomain Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB946T-100UG
This item is not returnable.
View return policy
Description
ADAM10 Monoclonal specifically detects ADAM10 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| ADAM10 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| O35598 | |
| ADAM10 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ADAM10 Thr215-Glu673 Accession # O35598 | |
| RUO | |
| 102 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 139712 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian, Kuzbanian protein homolog, MADM, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse ADAM10 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) rhADAM10 is observed and no cross-reactivity with rhADAM8, recombinant mouse (rm) ADAM9, rmADAM15, rhBACE, or rhTACE is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction