missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ADAM10 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB946A-100UG
This item is not returnable.
View return policy
Description
ADAM10 Monoclonal specifically detects ADAM10 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| ADAM10 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| a disintegrin and metalloprotease domain 10, a disintegrin and metalloproteinase domain 10, AD10, ADAM 10, ADAM metallopeptidase domain 10, CD156c, CD156c antigen, CDw156, disintegrin and metalloproteinase domain-containing protein 10, EC 3.4.24, HsT18717, kuz, Kuzbanian, Kuzbanian protein homolog, MADM, MADMEC 3.4.24.81, Mammalian disintegrin-metalloprotease | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse ADAM10 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) rhADAM10 is observed and no cross-reactivity with rhADAM8, recombinant mouse (rm) ADAM9, rmADAM15, rhBACE, or rhTACE is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 139712 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| O35598 | |
| ADAM10 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ADAM10, Thr215-Glu673, Accession No. O35598 | |
| 100 μg | |
| 102.0 | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction