missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse 5T4 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF5049-SP
This item is not returnable.
View return policy
Description
5T4 Polyclonal specifically detects 5T4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| 5T4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, M6P1, TPBG, trophoblast glycoprotein | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 7162 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| AAH58198 | |
| TPBG | |
| Mouse myeloma cell line NS0-derived recombinant mouse 5T4 Ser30-Ser361 Accession # AAH58198 | |
| 25 μg | |
| Primary | |
| Detects mouse 5T4 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human 5T4 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction