missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse 4-1BB Ligand/TNFSF9 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF1246
This item is not returnable.
View return policy
Description
4-1BB Ligand/TNFSF9 Polyclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot.
Specifications
| 4-1BB Ligand/TNFSF9 | |
| Polyclonal | |
| Unconjugated | |
| P41274 | |
| TNFSF9 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse 4-1BB Ligand/TNFSF9 Arg104-Glu309 Accession # P41274 | |
| 100 μg | |
| Primary | |
| Detects mouse 4-1BB Ligand/TNFSF9 in direct ELISAs and Western blots. In Western blots, approximatley 5% cross-reactivity with recombinant mouse (rm) Fas Ligand is observed and less than 1% cross-reactivity with rmOX40 Ligand, rmTWEAK, rmTRAIL, and rmTNF-alpha is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| 4-1BB ligand, 41BB Ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, TNFSF9, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8744 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction