Learn More
Leica Microsystems Mount Adapter
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
All systems include XY mechanical stage, coarse/fine focus, 4 position nosepiece, Auto off, two eyepieces 10x/20 (one fixed, one adjustable), dust cover, and immersion oil.
- The ideal choice for advanced college and university life science courses and professional training in medical, veterinary and dental schools
- EZStore™ design with integrated handle and cord wrap allows easy carrying, easy lifting and protection against microscope component damage
- The DM750 range feature LED illumination, eliminating the nuisance and expense of replacing bulbs
- Includes auto-off function that powers down the lighting after 2hr of unuse
- The optical system uses the same platform as Leica research grade microscopes
- Rackless stage system means there is no rack and pinion mechanism protruding from the side of the stage - preventing minor scratches or grazes to the skin or gloves
- Ultra hard wearing stage surface
- All systems include XY mechanical stage, course/fine focus, 4 position nosepiece, auto-off, two eyepieces 10x/20 (one fixed, one adjustable) dust cover and immersion oil
Order Info
Includes
Specifications
Specifications
| Type | Mount Adapter |
| Includes | C-mount Camera Adapter, DM500/750 |
| Quantity | 1 Ea. |