missing translation for 'onlineSavingsMsg'
Learn More

Motolimod, MedChemExpress

Product Code. 16453386
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453386

Brand: Medchem Express HY13773

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Motolimod (VTX-2337;VTX-378) is a selective Toll-like receptor 8 (TLR8) agonist, with an EC50 of approximately 100 nM.

Motolimod (VTX-2337

Specifications

Chemical Name or Material Motolimod
Physical Form Solid
CAS 926927-61-9
Color Light Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C28H34N4O2
For Use With (Application) Cancer-programmed cell death
Synonym VTX-2337 VTX-378
Solubility Information DMSO : 50 mg/mL (109.03 mM; Need ultrasonic)
SMILES O=C(C1=CC2=CC=C(C3=CC=C(C(N4CCCC4)=O)C=C3)C=C2N=C(N)C1)N(CCC)CCC
Molecular Weight (g/mol) 458.6
Formula Weight 458.6
Percent Purity 97.75%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.