missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motilin R/GPR38 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-09454-100UL
This item is not returnable.
View return policy
Description
Motilin R/GPR38 Polyclonal specifically detects Motilin R/GPR38 in Human samples. It is validated for Western Blot.
Specifications
| Motilin R/GPR38 | |
| Polyclonal | |
| Western Blot 1.0 ug/ml | |
| GPR38, motilin receptor, MTLR1 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human Motilin R/GPR38 (NP_001498). Peptide sequence ASINPILYNLISKKYRAAAFKLLLARKSRPRGFHRSRDTAGEVAGDTGGD | |
| 100 μg | |
| GPCR | |
| 2862 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS buffer, 2% sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction