missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motilin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97102-100ul
This item is not returnable.
View return policy
Description
Motilin Polyclonal antibody specifically detects Motilin in Human samples. It is validated for ELISA
Specifications
| Motilin | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| MGC138519, motilin, prepromotilin, promotilin | |
| Produced in rabbits immunized with purified, recombinant Human Motilin (Accession#: AAH69675.1; Met1-Lys115) | |
| 100 μg | |
| Signal Transduction | |
| 4295 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction