missing translation for 'onlineSavingsMsg'
Learn More

Motesanib Diphosphate, MedChemExpress

Product Code. 16433728
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16433728

Brand: Medchem Express HY10229

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Motesanib Diphosphate (AMG 706 Diphosphate) is a potent ATP-competitive inhibitor of VEGFR1/2/3 with IC50s of 2 nM/3 nM/6 nM, respectively, and has similar activity against Kit, and is approximately 10-fold more selective for VEGFR than PDGFR and Ret.

Motesanib Diphosphate (AMG 706 Diphosphate) is a potent ATP-competitive inhibitor of VEGFR1/2/3 with IC50s of 2 nM/3 nM/6 nM, respectively, and has similar activity against Kit, and is approximately 10-fold more selective for VEGFR than PDGFR and Ret.

Specifications

Chemical Name or Material Motesanib Diphosphate
CAS 857876-30-3
Color White
Purity Grade Notes Research
Molecular Formula C22H29N5O9P2
Quantity 10 mg
Synonym AMG 706 Diphosphate
Solubility Information DMSO : ≥ 110 mg/mL (193.17 mM) ∣H2O : 5 mg/mL (8.78 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1=CC=CN=C1NCC2=CC=NC=C2)NC3=CC(NCC4(C)C)=C4C=C3.O=P(O)(O)O.O=P(O)(O)O
Molecular Weight (g/mol) 569.44
Formula Weight 569.44
Percent Purity 99.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.