missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motesanib Diphosphate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Motesanib Diphosphate (AMG 706 Diphosphate) is a potent ATP-competitive inhibitor of VEGFR1/2/3 with IC50s of 2 nM/3 nM/6 nM, respectively, and has similar activity against Kit, and is approximately 10-fold more selective for VEGFR than PDGFR and Ret.
Specifications
Specifications
| Chemical Name or Material | Motesanib Diphosphate |
| CAS | 857876-30-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H29N5O9P2 |
| Quantity | 50 mg |
| Synonym | AMG 706 Diphosphate |
| Solubility Information | DMSO : ≥ 110 mg/mL (193.17 mM) ∣H2O : 5 mg/mL (8.78 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(C1=CC=CN=C1NCC2=CC=NC=C2)NC3=CC(NCC4(C)C)=C4C=C3.O=P(O)(O)O.O=P(O)(O)O |
| Molecular Weight (g/mol) | 569.44 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction