missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOSC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553276
This item is not returnable.
View return policy
Description
Immunogen sequence: FQVAYPDYCP LLIMTDASLV DLNTRMEKKM KMENFRPNIV VTGCDAFEED TWDELLIGSV EVKKVMACPR CILTTVDPDT GVIDRKQPLD TLKSYRL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
As a component of the benzamidoxime prodrug-converting complex required to reduce N-hydroxylated prodrugs, such as benzamidoxime. Also able to reduce N(omega)-hydroxy-L-arginine (NOHA) and N(omega)-hydroxy-N(delta)-methyl-L-arginine (NHAM) into L-arginine and N(delta)-methyl-L-arginine, respectively.
Specifications
| MOSC2 | |
| Polyclonal | |
| Unconjugated | |
| MARC2 | |
| 2810484M10Rik; mARC1; MARC2; Mg87; Mitochondrial amidoxime reducing component 2; moco sulfurase C-terminal domain-containing protein 2; MOCO sulphurase C-terminal domain containing 2; molybdenum cofactor sulfurase C-terminal domain-containing protein 2; MOSC domain-containing protein 2; MOSC domain-containing protein 2, mitochondrial; Mosc2; RP11-270A6.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54996 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969Z3 | |
| MARC2 | |
| Recombinant protein corresponding to Human MOSC2. Recombinant protein control fragment (Product #RP-90153). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction