missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MOSC2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA531602

Product Code. 13466127

  • £439.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: A549, HeLa. Predicted reactivity: Rhesus Monkey (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

As a component of the benzamidoxime prodrug-converting complex required to reduce N-hydroxylated prodrugs, such as benzamidoxime. Also able to reduce N(omega)-hydroxy-L-arginine (NOHA) and N(omega)-hydroxy-N(delta)-methyl-L-arginine (NHAM) into L-arginine and N(delta)-methyl-L-arginine, respectively.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MOSC2
Polyclonal
Unconjugated
MARC2
2810484M10Rik; mARC1; MARC2; Mg87; Mitochondrial amidoxime reducing component 2; moco sulfurase C-terminal domain-containing protein 2; MOCO sulphurase C-terminal domain containing 2; molybdenum cofactor sulfurase C-terminal domain-containing protein 2; MOSC domain-containing protein 2; MOSC domain-containing protein 2, mitochondrial; Mosc2; RP11-270A6.1
Rabbit
Antigen affinity chromatography
RUO
54996
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q969Z3
MARC2
Recombinant protein encompassing a sequence within the center region of human MOSC2. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.