missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOSC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
£178.00 - £456.00
Specifications
| Antigen | MOSC1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17760694
![]() |
Invitrogen™
PA5119290 |
200 μL |
£456.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17793164
|
Invitrogen™
PA5121771 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MOSC1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
MOSC1 (MOCO sulphurase C-terminal domain containing 1), also known as MARC1, is a 337 amino acid mitochondrial protein that is thought to function as an oxidoreductase. Existing as three alternatively spliced isoforms, MOSC1 contains one MOSC domain and binds molybdenum as a cofactor. The gene encoding MOSC1 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.Specifications
| MOSC1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat, Bovine | |
| MARC1 | |
| mARC1; mitochondrial amidoxime reducing component 1; mitochondrial amidoxime-reducing component 1; moco sulfurase C-terminal domain-containing protein 1; MOCO sulphurase C-terminal domain containing 1; molybdenum cofactor sulfurase C-terminal domain-containing protein 1; MOSC domain-containing protein 1; MOSC domain-containing protein 1, mitochondrial; MOSC1 | |
| MARC1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5VT66, Q9CW42 | |
| 537028, 64757, 66112, 690745 | |
| Recombinant protein MOCO Sulphurase C-Terminal Domain Containing Protein 1. The antigen corresponds to amino acid range 41-335 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title