missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOS Antibody (S3.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00
Specifications
| Antigen | MOS |
|---|---|
| Clone | S3.1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot |
Description
MOS Monoclonal specifically detects MOS in Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
| MOS | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Xenopus species | |
| 4342 | |
| Recombinant (Xenopus laevis) Mos fusion protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| S3.1 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| c-mos, EC 2.7.11, EC 2.7.11.1, MGC119962, MGC119963, MSV, oncogene MOS, Moloney murine sarcoma virus, Oocyte maturation factor mos, Proto-oncogene c-Mos, proto-oncogene serine/threonine-protein kinase mos, v-mos Moloney murine sarcoma viral oncogene homolog | |
| MOS | |
| IgG2a | |
| Protein A purified | |
| Able to immunoprecipitate MOS from cell/egg extracts. Cross-reacts with Xenopus laevis. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title