Learn More
Heinz Herenz Mortar Pressed Glass
Product Code. 30158472
Click to view available options
Material:
Glass
Unit Size:
Pack of 10
Product Code. 30158472
Brand: Heinz Herenz 1070582
Only null left
Add to basket
Add to basket
Mortar with 120 mm outer diameter. Pressed glass.
  • Mortar
  • 120 mm outer diameter
  • Pressed glass
Specifications
Finish Unglazed
Autoclavable Autoclavable
Product Type Mortar and Pestle
Material Glass
Type Mortar