missing translation for 'onlineSavingsMsg'
Learn More

Morphothiadin, MedChemExpress

Product Code. 16480577
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480577 50 mg 50mg
16453006 25 mg 25mg
16451119 1 mg 1mg
16446967 5 mg 5mg
16424248 10 mg 10mg
16405186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16480577 Supplier Medchem Express Supplier No. HY108917
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Morphothiadin is a potent inhibitor on the replication of both wild-type and adefovir-resistant HBV with an IC50 of 12 nM.

Morphothiadin is a potent inhibitor on the replication of both wild-type and adefovir-resistant HBV with an IC50 of 12 nM.

Specifications

Chemical Name or Material Morphothiadin
CAS 1092970-12-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H22BrFN4O3S
Quantity 50 mg
Synonym GLS4
Solubility Information DMSO : 62.5 mg/mL (122.70 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1=C(CN2CCOCC2)NC(C3=NC=CS3)=NC1C4=CC=C(F)C=C4Br)OCC
Molecular Weight (g/mol) 509.39
Formula Weight 509.39
Percent Purity 99.05%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.