missing translation for 'onlineSavingsMsg'
Learn More

Morpholino(4-piperidinyl)methanone hydrochloride, ≥90%, Thermo Scientific™

Catalog Number p-3076346
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
63214-57-3
C10H18N2O2
198.27
MFCD03372501
UVDVXQHUTBCRKT-UHFFFAOYSA-N
4-piperidine-4-carbonyl morpholine, morpholino piperidin-4-yl methanone, 4-piperidin-4-ylcarbonyl morpholine, morpholin-4-yl piperidin-4-yl methanone, 4-4-piperidinylcarbonyl morpholine, morpholin-4-yl-piperidin-4-yl-methanone, morpholin-4-yl 4-piperidyl ketone, 4-piperidin-4-yl carbonyl morpholine, methanone, 4-morpholinyl-4-piperidinyl
1520211
O=C(C1CCNCC1)N1CCOCC1
This item is not returnable. View return policy
63214-57-3
C10H18N2O2
198.27
MFCD03372501
UVDVXQHUTBCRKT-UHFFFAOYSA-N
4-piperidine-4-carbonyl morpholine, morpholino piperidin-4-yl methanone, 4-piperidin-4-ylcarbonyl morpholine, morpholin-4-yl piperidin-4-yl methanone, 4-4-piperidinylcarbonyl morpholine, morpholin-4-yl-piperidin-4-yl-methanone, morpholin-4-yl 4-piperidyl ketone, 4-piperidin-4-yl carbonyl morpholine, methanone, 4-morpholinyl-4-piperidinyl
1520211
O=C(C1CCNCC1)N1CCOCC1
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 4-(piperidine-4-carbonyl)morpholine
Formula Weight 234.73
Percent Purity 90%
Chemical Name or Material Morpholino(4-piperidinyl)methanone hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'