missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, Mikromol™

Product Code. 18022100
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18022100 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18022100 Supplier Mikromol Supplier No. MM0267.06

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Morpholine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Morpholine
CAS 110-91-8
Analyte or Component Names Morpholine
Shipping Condition Room Temperature
Molecular Formula C4 H9 N O
Certifications/Compliance ISO 9001
Synonym Morpholine, Molsidomine Imp. E (EP), Molsidomine Impurity E
InChI Formula InChI=1S/C4H9NO/c1-3-6-4-2-5-1/h5H,1-4H2
SMILES C1COCCN1
IUPAC Name morpholine
Molecular Weight (g/mol) 87.12
Quantity 100 mg
Formula Weight 87.0684 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.