missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, ACS, 99.0% min

Product Code. 11368307
Change view
Click to view available options
Quantity:
500 g
4 x 500 g
Unit Size:
2000g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368307 4 x 500 g 2000g
11378307 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368307 Supplier Thermo Scientific Alfa Aesar Supplier No. 031984.57
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-91-8 | C4H9NO | 87.12 g/mol

Morpholine is used to make other chemicals, as a corrosion inhibitor, and in detergents. It is also used as solvent and optical brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-91-8
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Specifications

Melting Point -5°C
Color Colorless
Density 1.007 g/mL
Boiling Point 129°C
Flash Point 35°C (95°F)
Odor Amine-like
Refractive Index 1.454
Quantity 4 x 500 g
UN Number UN2054
Beilstein 102549
Sensitivity Hygroscopic
Merck Index 14,6277
Solubility Information Soluble in water.
Formula Weight 87.12
Percent Purity ≤99.0%
Grade ACS
Physical Form Liquid
Chemical Name or Material Morpholine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.