missing translation for 'onlineSavingsMsg'
Learn More

Morpholine, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog Number 11378307
Click to view available options
Quantity:
500 g
4 x 500 g
110-91-8
C4H9NO
87.12
MFCD00005972
YNAVUWVOSKDBBP-UHFFFAOYSA-N
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
8083
CHEBI:34856
morpholine
C1COCCN1
110-91-8
C4H9NO
87.12
MFCD00005972
YNAVUWVOSKDBBP-UHFFFAOYSA-N
1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
8083
CHEBI:34856
morpholine
C1COCCN1

Morpholine is used to make other chemicals, as a corrosion inhibitor, and in detergents. It is also used as solvent and optical brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -5°C
Color Colorless
Density 1.007 g/mL
Boiling Point 129°C
Flash Point 35°C (95°F)
Odor Amine-like
Refractive Index 1.454
Quantity 500 g
UN Number UN2054
Beilstein 102549
Sensitivity Hygroscopic
Merck Index 14,6277
Solubility Information Soluble in water.
Formula Weight 87.12
Percent Purity ≤99.0%
Grade ACS
Physical Form Liquid
Chemical Name or Material Morpholine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'