missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Moroxydine Hydrochloride, Mikromol™
Description
Moroxydine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Moroxydine Hydrochloride |
| CAS | 3160-91-6 |
| Analyte or Component Names | Moroxydine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H13 N5 O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Moroxydine Hydrochloride, 4-Morpholinecarboximidamide, N-(aminoiminomethyl)-, hydrochloride (1:1), 4-Morpholinecarboxamidine, N-amidino-, monohydrochloride (8CI), 4-Morpholinecarboximidamide, N-(aminoiminomethyl)-, monohydrochloride (9CI), Guanidine, 4-morpholinecarboximidoyl-, hydrochloride (6CI), 4-(Amidinoamidino)morpholine hydrochloride, ABOB hydrochloride, Flumidin, Malinguchlorate, Miejunging, Moroxydine hydrochloride, Morpholinobiguanide hydrochloride, Morpholinobiguanide monohydrochloride, Spenitol, Virusmin, Virustat, N1,N1-Anhydrobis(beta-hydroxyethyl)biguanide hydrochloride |
| InChI Formula | InChI=1S/C6H13N5O.ClH/c7-5(8)10-6(9)11-1-3-12-4-2-11;/h1-4H2,(H5,7,8,9,10);1H |
| SMILES | Cl.NC(=N)NC(=N)N1CCOCC1 |
| IUPAC Name | N-carbamimidoylmorpholine-4-carboximidamide;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?