missing translation for 'onlineSavingsMsg'
Learn More

Moroxydine hydrochloride, MedChemExpress

Product Code. 16448296
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
10 g
Unit Size:
100mg
10g
1g
500mg
5g
This item is not returnable. View return policy

Product Code. 16448296

Brand: Medchem Express HYB0420A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Moroxydine hydrochloride (ABOB hydrochloride) is a synthetic antiviral compound chemically belonging to the series of the heterocyclic biguanidines.

Moroxydine hydrochloride (ABOB hydrochloride) is a synthetic antiviral compound chemically belonging to the series of the heterocyclic biguanidines.

Specifications

Chemical Name or Material Moroxydine hydrochloride
Physical Form Solid
CAS 3160-91-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C6H14ClN5O
For Use With (Application) COVID-19-anti-virus
Synonym ABOB hydrochloride
Solubility Information H2O : ≥ 100 mg/mL (481.56 mM) ∣DMSO : 50 mg/mL (240.78 mM; Need ultrasonic)
SMILES N=C(N1CCOCC1)NC(N)=N.Cl
Molecular Weight (g/mol) 207.66
Formula Weight 207.66
Percent Purity 99.57%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.