missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MORN2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563493

Product Code. 15711272

  • £480.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MNGFGRLEHF SGAVYEGQFK DNMFHGLGTY TFPNGAKYTG NFNENRVEGE GEYTDIQGLE WSGNFHFTAA PDLKLK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 96%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MORN2
Polyclonal
Unconjugated
MORN2
BLOCK27; MOPT; MORN motif protein in testis; MORN repeat containing 2; MORN repeat-containing protein 2; MORN2; protein containing single MORN motif in testis
Rabbit
Antigen Affinity Chromatography
RUO
729967
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q502X0
MORN2
Recombinant protein corresponding to Human MORN2. Recombinant protein control fragment (Product #RP-99706).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.