missing translation for 'onlineSavingsMsg'
Learn More

Moricizine, TRC

Product Code. 30468580
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468580 10 mg 10mg
30475650 50 mg 50mg
30475348 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30468580 Supplier Toronto Research Chemicals Supplier No. TRCM63502510MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31883-05-3
Molecular Formula C22H25N3O4S
Molecular Weight (g/mol) 427.52
Synonym N-[10-[3-(4-morpholinyl)-1-oxopropyl]-10H-phenothiazin-2-yl]Carbamic Acid Ethyl Ester, 10-(3-Morpholinopropionyl)phenothiazine-2-carbamic Acid Ethyl Ester, Ethyl 10-(beta-N-Morpholinylpropionyl)phenothiazine-2-carbamate, Ethyl 10-(beta-Morpholinopropionyl)phenothiazine-2-carbamate, G 214, Moracizine
IUPAC Name ethyl N-[10-(3-morpholin-4-ylpropanoyl)phenothiazin-2-yl]carbamate
SMILES CCOC(=O)Nc1ccc2Sc3ccccc3N(C(=O)CCN4CCOCC4)c2c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H25N3O4S/c1-2-29-22(27)23-16-7-8-20-18(15-16)25(17-5-3-4-6-19(17)30-20)21(26)9-10-24-11-13-28-14-12-24/h3-8,15H,2,9-14H2,1H3,(H,23,27)
Formula Weight 427.1566
Chemical Name or Material Moricizine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.