missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRG15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566444
This item is not returnable.
View return policy
Beskrivning
Immunogen sequence: QKANQEQYAEG KMRGAAPGKK TSGLQQKNVE VKTKKNKQK Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
MRG15 ia a component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. This modification may both alter nucleosome-DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis and DNA repair.
Specifikationer
| MRG15 | |
| Polyclonal | |
| Unconjugated | |
| MORF4L1 | |
| Eaf3; Esa1p-associated factor 3 homolog; FWP006; HSPC008; HSPC061; HsT17725; MEAF3; MGC10631; mKIAA4002; Morf4l1; MORF-related gene 15 protein; MORF-related gene on chromosome 15; MORFRG15; mortality factor 4 like 1; mortality factor 4-like protein 1; MRG15; PP368; Protein MSL3-1; S863-6; TEG-189; testis expressed gene 189; testis-expressed gene 189 protein; Tex189; transcription factor-like protein MRG15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10933 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBU8 | |
| MORF4L1 | |
| Recombinant Human MORF4. Recombinant protein control fragment (Product #RP-105217). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering