missing translation for 'onlineSavingsMsg'
Learn More

Mordant Yellow 10 (Technical Grade), TRC

Product Code. 30179463
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179463 1 g 1g
30179269 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179463 Supplier Toronto Research Chemicals Supplier No. TRCM6350001G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6054-99-5
Molecular Formula C13 H8 N2 O6 S . 2 Na
Molecular Weight (g/mol) 366.26
Synonym 2-Hydroxy-5-[2-(4-sulfophenyl)diazenyl]benzoic Acid Disodium Salt, 2-Hydroxy-5-[(4-sulfophenyl)azo]benzoic Acid Disodium Salt, C.I. Mordant Yellow 10 Disodium salt, Acid Chrome Yellow, Acid Mordant Dark Yellow GG, Acid Mordant Yellow 2G, Acidic Medium Dark Yellow GG, C.I. 14010, Chrome Yellow 2J, Chrome Yellow DF, Chrome Yellow K, Cromal Yellow S, Diacromo Yellow 2G, Durochrome Yellow 2G, Eniacromo Yellow GR, Fast Chrome Yellow GD, Hispacrom Yellow 2GS, Kenachrome Yellow MD, Lighthouse Chrome Yellow MD, MY 10, Magracrom Yellow, Mordant Yellow 10, Mordant Yellow GG, Ponsol Yellow SWR, Solochrome Yellow 2G, Solochrome Yellow 2GS, Superchrome Yellow 2G, Tertrochrome Yellow R, Tulachrome Yellow GG, Sulfasalazine Impurity I;
IUPAC Name disodium;2-hydroxy-5-[(E)-(4-sulfonatophenyl)diazenyl]benzoate
SMILES [Na+].[Na+].Oc1ccc(cc1C(=O)[O-])N=Nc2ccc(cc2)S(=O)(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H10N2O6S.2Na/c16-12-6-3-9(7-11(12)13(17)18)15-14-8-1-4-10(5-2-8)22(19,20)21;;/h1-7,16H,(H,17,18)(H,19,20,21);;/q;2*+1/p-2/b15-14+;;
Formula Weight 365.9898
Chemical Name or Material Mordant Yellow 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.