missing translation for 'onlineSavingsMsg'
Learn More

Mordant Black 11 (Technical Grade), TRC

Product Code. 30011510
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011510 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30011510 Supplier Toronto Research Chemicals Supplier No. TRCM63500310G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1787-61-7
Molecular Formula C20 H12 N3 O7 S . Na
Quantity 10 g
Synonym Mordant Black 11
InChI Formula InChI=1S/C20H13N3O7S.Na/c24-17-10-18(31(28,29)30)15-9-12(23(26)27)6-7-14(15)19(17)22-21-16-8-5-11-3-1-2-4-13(11)20(16)25;/h1-10,24-25H,(H,28,29,30);/q;+1/p-1/b22-21+;
SMILES [Na+].Oc1cc(c2cc(ccc2c1N=Nc3ccc4ccccc4c3O)[N+](=O)[O-])S(=O)(=O)[O-]
IUPAC Name sodium;3-hydroxy-4-[(E)-(1-hydroxynaphthalen-2-yl)diazenyl]-7-nitronaphthalene-1-sulfonate
Molecular Weight (g/mol) 461.38
Formula Weight 461.0294
Chemical Name or Material Eriochrome Black T
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.