missing translation for 'onlineSavingsMsg'
Learn More

MORC3 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18397235 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397235 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397235 Supplier Novus Biologicals Supplier No. NBP306255FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MORC3 Polyclonal specifically detects MORC3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORC3
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias KIAA0136, MORC family CW-type zinc finger 3, MORC family CW-type zinc finger protein 3, nuclear matrix protein NXP2, NXP2, ZCW5, ZCWCC3, Zinc finger CW-type coiled-coil domain protein 3, zinc finger, CW type with coiled-coil domain 3, zinc finger, CW-type with coiled-coil domain 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MORC3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23515
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.