missing translation for 'onlineSavingsMsg'
Learn More

MORC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18342662 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342662 20 μL 20µL
18312443 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18342662 Supplier Novus Biologicals Supplier No. NBP30512720UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MORC2 Polyclonal specifically detects MORC2 in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORC2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias AC004542.C22.1, KIAA0852, MORC family CW-type zinc finger 2, MORC family CW-type zinc finger protein 2, ZCW3, ZCWCC1, Zinc finger CW-type coiled-coil domain protein 1, zinc finger, CW type with coiled-coil domain 1, zinc finger, CW-type with coiled-coil domain 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-250 of human MORC2 (NP_055756.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 22880
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.