missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MORC2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA549339

Product Code. 15907334

  • £440.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MORC2 Polyclonal Antibody for Western Blot, ICC/IF

Exhibits a cytosolic function in lipogenesis, adipogenic differentiation, and lipid homeostasis by increasing the activity of ACLY, possibly preventing its dephosphorylation. May act as a transcriptional repressor. Down-regulates CA9 expression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MORC2
Polyclonal
Unconjugated
MORC2
8430403M08Rik; ATPase MORC2; ATPase MORC2A; CMT2Z; Kiaa0852; microrchidia 2A; MORC family CW-type zinc finger 2; MORC family CW-type zinc finger protein 2; MORC family CW-type zinc finger protein 2A; MORC2; Morc2a; ZCW3; ZCWCC1; zinc finger CW-type coiled-coil domain protein 1; zinc finger, CW-type with coiled-coil domain 1
Rabbit
Antigen affinity chromatography
RUO
22880
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9Y6X9
MORC2
KLH conjugated synthetic peptide between 828-862 amino acids from the C-terminal region of human MORC2.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.