missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MORC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551390
This item is not returnable.
View return policy
Description
Immunogen sequence: TSECLRIEPD TTALSTNHET IDLLVQILRN CLRYFLPPSF PISKKQLSAM NSDELISFPL KEYFKQYEVG LQNLCNSYQS RADSRAKASE ESLRTSERKL RETEEKLQKL RTNIVALLQK VQEDIDINTD DELDAYIED Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
Exhibits a cytosolic function in lipogenesis, adipogenic differentiation, and lipid homeostasis by increasing the activity of ACLY, possibly preventing its dephosphorylation. May act as a transcriptional repressor. Down-regulates CA9 expression.
Specifications
| MORC2 | |
| Polyclonal | |
| Unconjugated | |
| MORC2 | |
| 8430403M08Rik; ATPase MORC2; ATPase MORC2A; CMT2Z; Kiaa0852; microrchidia 2A; MORC family CW-type zinc finger 2; MORC family CW-type zinc finger protein 2; MORC family CW-type zinc finger protein 2A; MORC2; Morc2a; ZCW3; ZCWCC1; zinc finger CW-type coiled-coil domain protein 1; zinc finger, CW-type with coiled-coil domain 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22880 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6X9 | |
| MORC2 | |
| Recombinant protein corresponding to Human MORC2. Recombinant protein control fragment (Product #RP-91359). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction