Learn More
MORC2 Mouse, anti-Human, Clone: 1G12E7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68530-1-IG-150UL
This item is not returnable.
View return policy
Description
The MORC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MORC2. This antibody recognizes human antigen. The MORC2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| MORC2 | |
| 1G12E7 | |
| KIAA0852, MORC2, ZCW3, ZCWCC1 | |
| 150 μL | |
| 22880 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |