missing translation for 'onlineSavingsMsg'
Learn More

MORC2 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB600-236V

Product Code. 18465684

  • £492.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MORC2 Polyclonal antibody specifically detects MORC2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MORC2
Polyclonal
Western Blot
AC004542.C22.1, KIAA0852, MORC family CW-type zinc finger 2, MORC family CW-type zinc finger protein 2, ZCW3, ZCWCC1, Zinc finger CW-type coiled-coil domain protein 1, zinc finger, CW type with coiled-coil domain 1, zinc finger, CW-type with coiled-coil domain 1
A synthetic peptide, which maps to the region between residues 600-700 of human zinc finger, CW-type with coiled-coil domain 1 using the numbering given in entry NP_055756.1 (GeneID 22880).
0.1 mL
Transcription Factors and Regulators
22880
Store at 4°C in the dark.
IgG
Western Blot
DyLight 405
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.