missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOR106 Recombinant Mouse Monoclonal Antibody (MOR106)
Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554943
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
MOR106 is an anti-IL-17C monoclonal antibody that inhibits the activity of interleukin-17C, reducing inflammation.
Specifications
| MOR106 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA | |
| MOR106 | |
| Unconjugated | |
| IL17C; UNQ561/PRO1122 | |
| Recombinant Human IL17C. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction