missing translation for 'onlineSavingsMsg'
Learn More

MOPSO, 98%

Product Code. 11339628
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Unit Size:
10g
1kg
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339628 1 kg 1kg
11329628 10 g 10g
11319628 50 g 50g
11309628 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11339628 Supplier Thermo Scientific Alfa Aesar Supplier No. 044135.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biological buffers, pH range 6.2 to 7.6 | CAS: 68399-77-9 | C7H15NO5S | 225.259 g/mol

It is an important raw material used intermediate used in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals. Also used as biological buffers, pH range 6.2-7.6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68399-77-9
Molecular Formula C7H15NO5S
Molecular Weight (g/mol) 225.259
MDL Number MFCD00009629
InChI Key NUFBIAUZAMHTSP-UHFFFAOYSA-N
Synonym mopso, 2-hydroxy-3-morpholinopropanesulfonic acid, 2-hydroxy-3-morpholinopropane-1-sulfonic acid, 2-hydroxy-4-morpholinepropanesulphonic acid, 3-morpholino-2-hydroxypropanesulfonic acid, 3-n-morpholino-2-hydroxypropanesulfonic acid, 2-hydroxy-3-morpholin-4-yl propane-1-sulfonic acid, beta-hydroxy-4-morpholinepropanesulfonic acid, 3-4-morpholino-2-hydroxypropanesulfonic acid, 4-morpholinepropanesulfonic acid, .beta.-hydroxy
PubChem CID 109333
IUPAC Name 2-hydroxy-3-morpholin-4-ylpropane-1-sulfonic acid
SMILES C1COCCN1CC(CS(=O)(=O)O)O

Specifications

Melting Point 276°C to 279°C
Physical Form Powder
Quantity 1 kg
Solubility Information Water solubility under desired conditions ca.112,6g/L at 20°C.
Formula Weight 225.27
Percent Purity 98%
Chemical Name or Material MOPSO

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.