missing translation for 'onlineSavingsMsg'
Learn More

MOPS sodium salt, 97%

Product Code. 11473437
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11473437

Brand: Thermo Scientific Alfa Aesar A17214.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71119-22-7 | C7H14NNaO4S | 231.242 g/mol

3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

Solubility
Soluble in water (523 g/L at 20°C).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71119-22-7
Molecular Formula C7H14NNaO4S
MDL Number MFCD00064350
InChI Key MWEMXEWFLIDTSJ-UHFFFAOYSA-M
Synonym mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt
PubChem CID 3859613
IUPAC Name sodium;3-morpholin-4-ylpropane-1-sulfonate

Specifications

Melting Point ∼260°C (decomposition)
Quantity 500 g
Assay Percent Range 97%
Solubility Information Soluble aq. soln. (523 g/L (at 20°C).)
SMILES [Na+].[O-]S(=O)(=O)CCCN1CCOCC1
Molecular Weight (g/mol) 231.24
Formula Weight 231.25
Chemical Name or Material MOPS sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.