missing translation for 'onlineSavingsMsg'
Learn More

MOPS sodium salt, 98%, Thermo Scientific Chemicals

Catalog Number 11463437
Click to view available options
Quantity:
25 g
100 g
500 g
71119-22-7
C7H14NNaO4S
MFCD00064350
MWEMXEWFLIDTSJ-UHFFFAOYSA-M
mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt
3859613
sodium;3-morpholin-4-ylpropane-1-sulfonate
71119-22-7
C7H14NNaO4S
MFCD00064350
MWEMXEWFLIDTSJ-UHFFFAOYSA-M
mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt
3859613
sodium;3-morpholin-4-ylpropane-1-sulfonate

3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(N-Morpholino)propanesulfonic Acid, Na; an analog to MES used in polyacrylamide gel electrophoresis.

Solubility
Soluble in water (523 g/L at 20°C).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point ∽260°C (decomposition)
Quantity 100 g
Assay Percent Range 98%
Solubility Information Soluble in water (523g/L at 20°C).
SMILES C1COCCN1CCCS(=O)(=O)[O-].[Na+]
Molecular Weight (g/mol) 231.242
Formula Weight 231.25
Chemical Name or Material MOPS sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'