Learn More
Melford MOPS, Sodium Salt
Description
MOPS Sodium Salt, also known as 3-(N-Morpholino) propanesulfonic acid sodium salt, is a chemical compound that plays a significant role in the life science marketplace.
It is particularly effective in the pH range of 6.5 to 7.9, making it suitable for applications requiring slightly acidic to neutral pH conditions. MOPS Sodium Salt is commonly used in protein purification, enzyme assays, nucleic acid research, and electrophoresis techniques where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications.
One of the significant advantages of MOPS Sodium Salt is its compatibility with biological systems and enzymes. It exhibits minimal interference with enzymatic activity and other biological processes, ensuring reliable and accurate results.
Specifications
Specifications
| Chemical Name or Material | MOPS, Sodium Salt |
| CAS | 71119-22-7 |
| Quantity | 500 g |
| Molecular Formula | C7H14NO4SNa |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.