missing translation for 'onlineSavingsMsg'
Learn More

Melford MOPS, Sodium Salt

Product Code. 30018538
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Product Code. Quantity unitSize
30018538 1 kg 1kg
30018539 500 g 500g
30018537 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 30018538

Brand: Melford M920401000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MOPS Sodium Salt, also known as 3-(N-Morpholino) propanesulfonic acid sodium salt, is a chemical compound that plays a significant role in the life science marketplace.

MOPS Sodium Salt, also known as 3-(N-Morpholino) propanesulfonic acid sodium salt, is a chemical compound that plays a significant role in the life science marketplace. It is a zwitterionic buffer commonly used in biological and biochemical research and is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures.

It is particularly effective in the pH range of 6.5 to 7.9, making it suitable for applications requiring slightly acidic to neutral pH conditions. MOPS Sodium Salt is commonly used in protein purification, enzyme assays, nucleic acid research, and electrophoresis techniques where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications.

One of the significant advantages of MOPS Sodium Salt is its compatibility with biological systems and enzymes. It exhibits minimal interference with enzymatic activity and other biological processes, ensuring reliable and accurate results.

Specifications

Chemical Name or Material MOPS, Sodium Salt
CAS 71119-22-7
Quantity 1 kg
Molecular Formula C7H14NO4SNa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.