missing translation for 'onlineSavingsMsg'
Learn More

Hoefer MOPS-SDS Running Buffer

Product Code. 30286585
Change view
Click to view available options
Quantity:
10
Unit Size:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30286585 10 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30286585 Supplier Hoefer Supplier No. MSGB10
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Hoefer MOPS-SDS running buffer is ideal companion with Hoefer precast gels to ensure a perfect electrophoresis result.

  • Specification:
  • System:MOPS SDS
  • Form: white powder
  • Package:10/pack
  • Weight:0.5 lb.

Specifications

Chemical Name or Material Buffer powder
For Use With (Application) Eletrophoresis experiment
Physical Form Powder
Quantity 10
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.