missing translation for 'onlineSavingsMsg'
Learn More

Melford MOPS, Free Acid

Product Code. 30018536
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
250g
25g
500g
5kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018536 5 kg 5kg
30018533 25 g 25g
30018531 100 g 100g
30018535 500 g 500g
30018532 1 kg 1kg
30018534 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30018536 Supplier Melford Supplier No. M920205000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MOPS, or 3-(N-Morpholino) propanesulfonic acid, is a zwitterionic buffer.

MOPS, or 3-(N-Morpholino) propanesulfonic acid, is a zwitterionic buffer. MOPS is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures. It is particularly effective in the pH range of 6.5 to 7.9, making it suitable for applications requiring slightly acidic to neutral pH conditions.

MOPS is commonly used in protein purification, enzyme assays, nucleic acid research, and electrophoresis techniques where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications.

One of the significant advantages of MOPS is its compatibility with biological systems and enzymes. It exhibits minimal interference with enzymatic activity and other biological processes, ensuring reliable and accurate results.

Heavy Metals: <5 ppm

pKa: (20°C) 7.20 ± 0.2

pH Range: 6.5 to 7.9

Specifications

Chemical Name or Material MOPS, Free Acid
CAS 1132-61-2
Quantity 5 kg
Molecular Formula C7H15NO4S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.