missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MOPS, 99%, for biochemistry

Product Code. 10747573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10747573 500 g Plastic bottle 500g
10644862 25 g Plastic bottle 25g
10245560 100 g Plastic bottle 100g
10765052 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10747573 Supplier Thermo Scientific Chemicals Supplier No. 172635000
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-Morpholino)propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Absorbance 0.03 max. at 280nm
Absorbance Conditions (1M aq. soln.) (1 cm cell)
Melting Point 277.0°C to 282.0°C
CAS Min % 98.5
CAS Max % 100.0
Color White
Decomposition Information >277°C
pH 3.0 to 4.5 (1% aq. soln. at 25°C)
Physical Form Crystalline Powder
Quantity 500 g
Flash Point 116°C
Assay Percent Range 99%
Infrared Spectrum Authentic
Packaging Plastic bottle
Merck Index 15, 6352
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: <10g/L ethanol (20°C)
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
Formula Weight 209.26
Percent Purity 99%
Grade Biochemical
Chemical Name or Material MOPS, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.