missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals MOPS, 99%, for biochemistry

Product Code. 10245560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
Product Code. Quantity Packaging unitSize
10245560 100 g Plastic bottle 100g
10644862 25 g Plastic bottle 25g
10747573 500 g Plastic bottle 500g
10765052 2.5 kg Plastic bottle 2.5kg
4 options
This item is not returnable. View return policy

Product Code. 10245560

Brand: Thermo Scientific Chemicals 172631000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-Morpholino)propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Absorbance (1 M aq. soln.) (1 cm cell)
Absorbance Conditions (1M aq. soln.) (1 cm cell)
CAS Min % 98.5
CAS Max % 100.0
Color White
Decomposition Information >277°C
pH 3.0 to 4.5 (1% aq. soln. at 25°C)
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Packaging Plastic bottle
Merck Index 15, 6352
IUPAC Name 4-(3-sulfonatopropyl)morpholin-4-ium
Formula Weight 209.26
Percent Purity ≥98.5%
Grade Biochemical
Chemical Name or Material MOPS, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.