missing translation for 'onlineSavingsMsg'
Learn More

MOPS 10X Solution, Fisher BioReagents™

Product Code. 11889191
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 11889191

Brand: Fisher BioReagents BP29001

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Premixed MOPS buffer solution (pH = 7.0) at 10X concentration. A low ionic strength buffer used for electrophoretic separation of formaldehyde denatured RNA

A low ionic strength buffer used for electrophoretic separation of RNA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2, 7732-18-5, 1310-73-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-morpholinopropane-1-sulfonic acid, morpholinopropanesulfonic acid, morpholinopropane sulfonic acid, 4-morpholinepropane sulfonic acid, unii-273bp63nv3, 3-morpholinopropanesulfonic acid, 3-n-morpholinopropanesulfonic acid, 3-n-morpholino propanesulfonic acid, 4-morpholinepropanesulfonic acid, 3-[N-Morpholino]propane sulfonic Acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 4-(3-sulfonatopropyl)morpholin-4-ium
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Physical Form Liquid
Quantity 1 L
DNase DNase free
Filtered Through 0.2-micron filtered
Packaging Nalgene Poly Bottle
Protease Protease free
Merck Index 15, 6352
Concentration or Composition (by Analyte or Components) 10X solution contains 0.2M MOPS, 20mM sodium acetate trihydrate, 10mM disodium EDTA and NaOH to pH of 7.0
Chemical Name or Material MOPS, 10X Solution, pH 7
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.