missing translation for 'onlineSavingsMsg'
Learn More

Moperone, TRC

Product Code. 30135872
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135872 25 mg 25mg
30133917 5 mg 5mg
30005898 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135872 Supplier Toronto Research Chemicals Supplier No. TRCM61050025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1050-79-9
Molecular Formula C22 H26 F N O2
Molecular Weight (g/mol) 355.45
Synonym Butyrophenone, 4'-fluoro-4-(4-hydroxy-4-p-tolylpiperidino)- (6CI,7CI,8CI), 1-(4-Fluorophenyl)-4-[4-hydroxy-4-(4-methylphenyl)-1-piperidinyl]-1-butanone, 1-(3'-p-Fluorobenzoylpropyl)-4-hydroxy-4-p-tolylpiperidine, 4'-Fluoro-4-(4-hydroxy-4-p-tolylpiperidino)butyrophenone, Luvatren, Luvatrena, Meperon, Methylperidol, Moperone, p-Fluoro-4-(4'-hydroxy-4'-p-methylphenylpiperidino)butyrophenone, p-Fluoro-4-(4'-hydroxy-4'-p-tolylpiperidino)butyrophenone, ω-(4-Hydroxy-4-p-tolylpiperidino)-p-fluorobutyrophenone
IUPAC Name 1-(4-fluorophenyl)-4-[4-hydroxy-4-(4-methylphenyl)piperidin-1-yl]butan-1-one
SMILES Cc1ccc(cc1)C2(O)CCN(CCCC(=O)c3ccc(F)cc3)CC2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H26FNO2/c1-17-4-8-19(9-5-17)22(26)12-15-24(16-13-22)14-2-3-21(25)18-6-10-20(23)11-7-18/h4-11,26H,2-3,12-16H2,1H3
Formula Weight 355.1948
Chemical Name or Material Moperone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.