missing translation for 'onlineSavingsMsg'
Learn More

Montmorillonite, naturally occurring mineral, Thermo Scientific™

Product Code. p-7122658
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329118 25 g 25g
11319118 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329118 Supplier Thermo Scientific Alfa Aesar Supplier No. 042531.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used in industrial chromatographic techniques, widely used in petroleum industry, catalyst carrier. It acts as a solid Lewis acid catalyst.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1318-93-0
Molecular Formula Al2H2O12Si4
Molecular Weight (g/mol) 360.307
MDL Number MFCD00132796
InChI Key GUJOJGAPFQRJSV-UHFFFAOYSA-N
Synonym montmorillonite, bentolite, gelwhite l, montmorillonite ksf, montmorillonite k 10, unii-a585mn1h2l, envirobent, neokunibond, arcillite, bedelix
PubChem CID 71586775
IUPAC Name dialuminum;dioxosilane;oxygen(2-);hydrate
SMILES O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Odor Odorless
Chemical Name or Material Montmorillonite
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.