missing translation for 'onlineSavingsMsg'
Learn More

Montmorillonite K-10, granules

Product Code. 15397390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Unit Size:
100g
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15397390 5 kg 5kg
15250309 100 g 100g
15387390 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15397390 Supplier Thermo Scientific Acros Supplier No. 456070050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1318-93-0 | Al2H2O12Si4 | 360.307 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1318-93-0
Molecular Formula Al2H2O12Si4
Molecular Weight (g/mol) 360.307
InChI Key GUJOJGAPFQRJSV-UHFFFAOYSA-N
Synonym montmorillonite, bentolite, gelwhite l, montmorillonite ksf, montmorillonite k 10, unii-a585mn1h2l, envirobent, neokunibond, arcillite, bedelix
PubChem CID 71586775
IUPAC Name dialuminum;dioxosilane;oxygen(2-);hydrate
SMILES O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]

Specifications

Quantity 5 kg
Fieser 15,213
Merck Index 11,6168
Density 0.500 to 0.600 kg/l
Chemical Name or Material Montmorillonite K-10

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.