missing translation for 'onlineSavingsMsg'
Learn More

Montelukast sodium, MedChemExpress

Product Code. 16426166
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16426166

Brand: Medchem Express HY13315

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Montelukast sodium (MK0476) is a potent, selective and orally active antagonist of cysteinyl leukotriene receptor 1 (CysLT1).

Montelukast sodium can be used for the reseach of asthma and liver injury. Montelukast sodium also has an antioxidant effect in intestinal ischemia-reperfusion injury, and could reduce cardiac damage. Montelukast sodium decreases eosinophil infiltration into the asthmatic airways. Montelukast sodium can also be used for COVID-19 research.

Specifications

Chemical Name or Material Montelukast sodium
Physical Form Solid
CAS 151767-02-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C35H35ClNNaO3S
For Use With (Application) COVID-19-immunoregulation
Synonym MK0476
Solubility Information DMSO : 50 mg/mL (82.21 mM; Need ultrasonic)
SMILES ClC1=CC2=C(C=C1)C=CC(/C=C/C3=CC([C@H](SCC4(CC(O[Na])=O)CC4)CCC5=CC=CC=C5C(C)(O)C)=CC=C3)=N2
Molecular Weight (g/mol) 608.17
Formula Weight 608.17
Percent Purity 98.62%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.